Learn More
ABclonal Technology ORP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A93141000UL
Encompass_Preferred
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist, they encode different isoforms.
Catalog No. 50-001-34645