Learn More
ABclonal Technology NCTR2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9320500UL
Encompass_Preferred
Predicted to enable signaling receptor activity. Predicted to be involved in cellular defense response and signal transduction. Predicted to be located in plasma membrane. Predicted to be integral component of plasma membrane. Predicted to be active in cell surface.
Catalog No. 50-001-34752