Learn More
ABclonal Technology CAP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A93301000UL
Encompass_Preferred
The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. The human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-34724