Learn More
ABclonal Technology HOOK2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9337500UL
Encompass_Preferred
Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.
Catalog No. 50-001-34778