Learn More
ABclonal Technology PPM1E Rabbit mAb
SDP
Supplier:  ABclonal Technology A9374500UL
Encompass_Preferred
This gene encodes a member of the PPM family of serine/threonine-protein phosphatases. The encoded protein is localized to the nucleus and dephosphorylates and inactivates multiple substrates including serine/threonine-protein kinase PAK 1, 5-AMP-activated protein kinase (AMPK) and the multifunctional calcium/calmodulin-dependent protein kinases. Alternatively spliced transcript variants have been observed for this gene.
Catalog No. 50-001-36406