Learn More
ABclonal Technology MYL12B Rabbit mAb
SDP
Supplier:  ABclonal Technology A9387500UL
Encompass_Preferred
The activity of nonmuscle myosin II (see MYH9, MIM 160775) is regulated by phosphorylation of a regulatory light chain, such as MRLC2. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments (Iwasaki et al., 2001 [PubMed 11942626]).
Catalog No. 50-001-34646