Learn More
ABclonal Technology CNAP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A93911000UL
Encompass_Preferred
Enables histone binding activity. Involved in mitotic chromosome condensation. Located in condensed chromosome, cytosol, and nucleoplasm. Part of condensin complex. Colocalizes with cytoplasm and nuclear chromosome. Implicated in primary autosomal recessive microcephaly.
Catalog No. 50-001-34825