Learn More
ABclonal Technology MMUT Rabbit mAb
SDP
Supplier:  ABclonal Technology A94061000UL
Encompass_Preferred
This gene encodes the mitochondrial enzyme methylmalonyl Coenzyme A mutase. In humans, the product of this gene is a vitamin B12-dependent enzyme which catalyzes the isomerization of methylmalonyl-CoA to succinyl-CoA, while in other species this enzyme may have different functions. Mutations in this gene may lead to various types of methylmalonic aciduria.
Catalog No. 50-001-35597