Learn More
ABclonal Technology Calumenin Rabbit mAb
SDP
Supplier:  ABclonal Technology A9416500UL
Encompass_Preferred
The product of this gene is a calcium-binding protein localized in the endoplasmic reticulum (ER) and it is involved in such ER functions as protein folding and sorting. This protein belongs to a family of multiple EF-hand proteins (CERC) that include reticulocalbin, ERC-55, and Cab45 and the product of this gene. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-34833