Learn More
ABclonal Technology PCBD1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A94261000UL
Encompass_Preferred
This gene encodes a member of the pterin-4-alpha-carbinolamine dehydratase family. The encoded protein has been identified as a moonlighting protein based on its ability to perform mechanistically distinct functions. The encoded protein functions as both a dehydratase involved in tetrahydrobiopterin biosynthesis, and as a cofactor for HNF1A-dependent transcription. A deficiency of this enzyme leads to hyperphenylalaninemia. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-36935