Learn More
Cayman Chemical A943931hydroCLOrID hydra 10mg
SDP
Supplier:  Cayman Chemical 3455410 MG
Encompass_Preferred
A histamine H4 receptor antagonist (Ki = 3.8 nM); inhibits clobenpropit-induced scratching in mice (ED50 = 26 µmoles/kg, i.p.)
Catalog No. 50-294-7466