Learn More
ABclonal Technology WSTF Rabbit mAb
SDP
Supplier:  ABclonal Technology A9519500UL
Encompass_Preferred
This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
Catalog No. 50-001-35415