Learn More
ABclonal Technology Cdx1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9546500UL
Encompass_Preferred
This gene is a member of the caudal-related homeobox transcription factor gene family. The encoded DNA-binding protein regulates intestine-specific gene expression and enterocyte differentiation. It has been shown to induce expression of the intestinal alkaline phosphatase gene, and inhibit beta-catenin/T-cell factor transcriptional activity.
Catalog No. 50-001-36181