Learn More
ABclonal Technology HIPK2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9552500UL
Encompass_Preferred
This gene encodes a conserved serine/threonine kinase that is a member of the homeodomain-interacting protein kinase family. The encoded protein interacts with homeodomain transcription factors and many other transcription factors such as p53, and can function as both a corepressor and a coactivator depending on the transcription factor and its subcellular localization. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-36217