Learn More
ABclonal Technology Hsc70-interacting protein (HIP) Rabbit mAb
SDP
Supplier:  ABclonal Technology A95671000UL
Encompass_Preferred
The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that it is a candidate tumor suppressor gene. Alternative splicing results in multiple transcript variants encoding different isoforms.
Catalog No. 50-001-36992