Learn More
ABclonal Technology GABARAPL2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9585500UL
Encompass_Preferred
Enables ubiquitin protein ligase binding activity. Involved in negative regulation of proteasomal protein catabolic process and protein localization to endoplasmic reticulum. Located in Golgi membrane and autophagosome membrane.
Catalog No. 50-001-35501