Learn More
ABclonal Technology PSMD14 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9608500UL
Encompass_Preferred
This gene encodes a component of the 26S proteasome. The 26S proteasome is a large multiprotein complex that catalyzes the degradation of ubiquitinated intracellular proteins. The encoded protein is a component of the 19S regulatory cap complex of the 26S proteasome and mediates substrate deubiquitination. A pseudogene of this gene is also located on the long arm of chromosome 2.
Catalog No. 50-001-35521