Learn More
ABclonal Technology SFRP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A96561000UL
Encompass_Preferred
This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. Members of this family act as soluble modulators of Wnt signaling, epigenetic silencing of SFRP genes leads to deregulated activation of the Wnt-pathway which is associated with cancer. This gene may also be involved in determining the polarity of photoreceptor cells in the retina.
Catalog No. 50-001-35680