Learn More
ABclonal Technology γ-Tubulin Rabbit mAb
SDP
Supplier:  ABclonal Technology A96571000UL
Encompass_Preferred
This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7.
Catalog No. 50-001-34794