Learn More
ABclonal Technology TOMM22 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9666500UL
Encompass_Preferred
The protein encoded by this gene is an integral membrane protein of the mitochondrial outer membrane. The encoded protein interacts with TOMM20 and TOMM40, and forms a complex with several other proteins to import cytosolic preproteins into the mitochondrion.
Catalog No. 50-001-35545