Learn More
ABclonal Technology Parathyroid Hormone (PTH) Rabbit mAb
SDP
Supplier:  ABclonal Technology A97041000UL
Encompass_Preferred
This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-35489