Learn More
ABclonal Technology RAP1A + RAP1B Rabbit mAb
SDP
Supplier:  ABclonal Technology A97251000UL
Encompass_Preferred
This gene encodes a member of the RAS-like small GTP-binding protein superfamily. Members of this family regulate multiple cellular processes including cell adhesion and growth and differentiation. This protein localizes to cellular membranes and has been shown to regulate integrin-mediated cell signaling. This protein also plays a role in regulating outside-in signaling in platelets. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 5, 6 and 9. [provided by RefSeq, Oct 2011]
Catalog No. 50-001-35671