Learn More
ABclonal Technology IQGAP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A97651000UL
Encompass_Preferred
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
Catalog No. 50-001-37079