Learn More
ABclonal Technology USP9X Rabbit mAb
SDP
Supplier:  ABclonal Technology A9782500UL
Encompass_Preferred
This gene is a member of the peptidase C19 family and encodes a protein that is similar to ubiquitin-specific proteases. Though this gene is located on the X chromosome, it escapes X-inactivation. Mutations in this gene have been associated with Turner syndrome. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Catalog No. 50-001-35623