Learn More
Selleck Chemical LLC 2-O-a-D-Glucopyranosyl-L-ascorbic acid S3215-25mg
SDP
Supplier:  Selleck Chemical LLC S321525MG
Encompass_Preferred
2-O- -D-Glucopyranosyl-l-ascorbic acid (AA-2G) which is a glucoside derivative of ascorbic acid (AA) shows vitamin C activity after enzymatic hydrolysis to ascorbic acid The antitumor activity of AA-2G is caused by ROS generated by AA released by rapid hydrolysis of AA-2G
Catalog No. 50-310-8569