Learn More
Medchemexpress LLC Aa-t3a-c12 | 2938207-23-7 | 99.76% | 1059.72 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14885910MM/1ML
Encompass_Preferred
AA-T3A-C12 is an anisamide ligand-tethered lipidoid (AA-lipidoid) that mediates great RNA delivery and transfection of activated fibroblasts. It is for research use only.
  • Mediates great RNA delivery and transfection of activated fibroblasts.
  • Enhances uptake of siRNA LNP by TGF-β-stimulated 3T3 fibroblasts.
  • Enables robust gene knockdown in activated fibroblasts.
  • Significantly down-regulates HSP47 expression in fibrotic mice.
Catalog No. 50-003-61737