Learn More
Medchemexpress LLC AA-T3A-C12 | 2938207-23-7 | 99.8% | 1059.72 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY148859100MG
Encompass_Preferred
AA-T3A-C12 is an anisamide ligand-tethered lipidoid (AA-lipidoid) that mediates excellent RNA delivery and transfection of activated fibroblasts. It is for research use only.
  • Enhances uptake of siRNA LNP by TGF-β-stimulated 3T3 fibroblasts
  • Enables robust gene knockdown in activated fibroblasts
  • Significantly down-regulates HSP47 expression in fibrotic mice
  • Appearance: Liquid, colorless to light yellow
Catalog No. 50-003-59546