Learn More
Selleck Chemical LLC AA147-E0745-5MG
SDP
Supplier:  Selleck Chemical LLC E07455MG
Encompass_Preferred
AA147 a small molecule endoplasmic reticulum (ER) proteostasis regulator selectively activates ATF6 arm of the unfolded protein response (UPR) extracted from patent WO2017117430A1 compound 147
Catalog No. 50-313-4433