Learn More
Medchemexpress LLC HY-18522 5mg Medchemexpress, AA26-9 CAS:1312782-34-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY185225MG
Encompass_Preferred
Medchemexpress, HY-18522 5mg AA26-9 CAS:1312782-34-5 AA26-9 is a potent and broad spectrum serine hydrolase inhibitor. AA26-9 targets included serine peptidases, lipases, amidases, esterases, and thioesterases. AA26-9 shows inhibitory activity against approximately 1/3 of the 40+ serine hydrolases detected in immortalized T cell lines . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2044