Learn More
ACROBiosystems Biotinylated Human HLA-A*0201 Glycipan 3 (FVGEFFTDV) Complex Protein (HLG-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*0201) & FVGEFFTDV & Ile 21 - Met 119 (B2M) (Accession # A0A140T913 (HLA-A*02
Supplier:  ACROBiosystems HLGH82E425UG
Encompass_Preferred
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
Catalog No. 50-225-4100