Learn More
Norgen Biotek Corp AAV Purification Maxi Slurry Kit, 1-10 preps, Spin column chromatography, Cell pellet/culture media/mixed input, 45 mL - 90 mL input volume per column, Up to 900 mL total input volume, 2.5 - 3.5 hours preparation time, 1 mL - 10 mL minimum elution volume, Up to 90% AAV recovery
Supplier:  Norgen Biotek Corp 63250
Encompass_Preferred
AAV Purification Maxi Slurry Kit, 1-10 preps, Spin column chromatography, Cell pellet/culture media/mixed input, 45 mL - 90 mL input volume per column, Up to 900 mL total input volume, 2.5 - 3.5 hours preparation time, 1 mL - 10 mL minimum elution volume, Up to 90% AAV recovery
Catalog No. NC3371952