Learn More
Cygnus Technologies AVIPure AAV2 Residual Ligand Assay Kit, Two-Site Immuno-Enzymatic ELISA, Less Than 1 ng/mL Sensitivity, Unit: Kit
Supplier:  Cygnus Technologies F1000
Encompass
The AVIPure – AAV2 Residual Ligand Assay Kit is a highly sensitive ELISA for quantifying residual Avitide AVIPure – AAV2 Ligand in AAV2 purification processes. It ensures safety and purity by accurately measuring ligand leached from chromatography media, supporting optimal purification and quality control. Unlike other kits, it employs a well-qualified sample treatment method, enabling detection of low concentrations, making it essential for research and manufacturing applications.
Catalog No. NC3310389