Learn More
Dojindo Molecular Technologies Inc Ab-10 Rapid R-Phycoerythrin Labeling Kit (3 samples) is used to prepare R-Phycoerythrin-labeled antibodies from 10 µg protein samples, Just mix antibody and reagent, recover conjugates within 30 min.
SDP
Supplier:  Dojindo Molecular Technologies Inc LK3410
Encompass_Preferred
R-PE is one of the phycobiliproteins. It emits a red fluorescence at around 578 nm and can be excited at 488nm, although the maximum excitation is 564 nm. Ab-10 Rapid R-Phycoerythrin Labeling Kit is used for the preparation of R-phycoerythrin-labeled IgG for immunostaining and the preparation of labeled cellular proteins for tracing. This kit can bind R-Phycoerythrin to 10 µg antibody in less than 30 minutes, and it contains all the necessary reagents for antibody labeling. The labeling agent included in the kit has a succinimidyl ester group, that can easily make a covalent bond with an amino group of the target antibody without any activation process. The procedure is very simple: just mix antibody and labeling agent. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/)
Catalog No. NC1960983