Learn More
Cayman Chemical AB-CHMINACA metabolIte M6 5mg
SDP
Supplier:  Cayman Chemical 164535 MG
Encompass_Preferred
A potential metabolite of AB-CHMINACA in which the isopropyl group has been oxidized; intended for research and forensic applications
Catalog No. 50-297-0257