Learn More
TARGETMOL CHEMICALS INC Abacavir 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1267100MG
Encompass_Preferred
Also available in 1 g, 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Abacavir (Ziagen) is a nucleoside reverse transcriptase inhibitor analog of guanosine. This agent decreases HIV viral loads, retards or prevents the damage to the immune system, and reduces the risk of developing AIDS. Purity 99.81%
Catalog No. 50-287-9826