Learn More
Medchemexpress LLC Abacavir | 136470-78-5 | 98.97% | 286.34 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1742350MG
Encompass_Preferred
Abacavir is an orally active and competitive nucleoside reverse transcriptase inhibitor. It inhibits the replication of HIV and shows anticancer activity in prostate cancer cell lines. Abacavir can also trespass the blood-brain-barrier and suppresses telomerase activity.
  • Orally active
  • Competitive nucleoside reverse transcriptase inhibitor
  • Inhibits HIV replication
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity
Catalog No. 50-003-86100