Learn More
Medchemexpress LLC Abacavir | 136470-78-5 | CB5698138 | 99.0% | 286.34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17423100MG
Encompass_Preferred
Abacavir is an orally active and competitive nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and shows anticancer activity in prostate cancer cell lines. Abacavir is capable of trespassing the blood-brain-barrier and suppressing telomerase activity.
  • Inhibits HIV replication
  • Exhibits anticancer activity in prostate cancer cell lines
  • Crosses the blood-brain-barrier
  • Suppresses telomerase activity
  • Induces fat apoptosis
  • Inhibits cell growth and affects cell cycle progression in prostate cancer cell lines
  • Reduces cell migration and inhibits cell invasion
Catalog No. 50-003-84028