Learn More
Medchemexpress LLC Abacavir hydroxyacetate | 1446418-48-9 | 99.6% | 344.37 | C16H20N6O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10664710MM 1ML
Encompass_Preferred
Abacavir hydroxyacetate is a research-grade small molecule reported as an orally active inhibitor of IL-20 and PRINS, used to investigate inflammatory skin responses and related immune pathways. It is supplied for laboratory research in solution and solid forms, with molecular formula C16H20N6O3 and molecular weight 344.37 g·mol-1.
  • Orally active IL-20 and PRINS inhibitor for inflammation research.
  • Available as a 10 mM solution in DMSO for ready-to-use experiments.
  • Also supplied as solid samples for custom formulation or storage.
  • High analytical purity reported to support reproducible results.
  • Molecular formula C16H20N6O3; molecular weight 344.37 g·mol-1.
Catalog No. 50-002-40073