Learn More
Selleck Chemical LLC Abacavir sulfate S3165-50mg
SDP
Supplier:  Selleck Chemical LLC S316550MG
Encompass_Preferred
Abacavir (1592U89) is a commonly used nucleoside analogue with potent antiviral activity against HIV-1
Catalog No. 50-311-0033