Learn More
Medchemexpress LLC Abafungin | 129639-79-8 | 99.1% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1198475MG
Encompass_Preferred
Abafungin is an antifungal research compound that inhibits transmethylation at the C-24 position of the sterol side chain catalyzed by sterol-C-24-methyltransferase. Provided as a high-purity solid for in vitro antifungal and biochemical studies. Not for human or clinical use.
  • High purity suitable for research applications.
  • Reported molecular weight 378.49 and formula C21H22N4OS.
  • Available in small pack sizes for screening and assay development.
  • Includes data sheet, certificate of analysis, and safety data sheet.
  • Acts as an inhibitor of sterol-C-24-methyltransferase.
Catalog No. 50-002-31889