Learn More
Selleck Chemical LLC Abaloparatide P1092-100MG
SDP
Supplier:  Selleck Chemical LLC P1092100MG
Encompass_Preferred
Abaloparatide (BA058 BIM-44058 ITM-058) is a novel 34-amino acid peptide selected to be a potent and selective activator of the parathyroid hormone receptor (PTH1R) signaling pathway with an IC50 of 0 117 nM in SOST analysis
Catalog No. 50-000-41032