Learn More
Medchemexpress LLC Abametapir | 1762-34-1 | 184.24 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW004546100G
Encompass_Preferred
Abametapir is a metalloproteinase (MMP) inhibitor that targets metalloproteinases crucial for egg hatching and louse development. It can inhibit the hatching of both head and body lice.
  • Targets metalloproteinases critical for egg hatching and louse development.
  • Inhibits the hatching of head and body lice.
  • Chelates heavy metal ions such as iron, copper, and zinc.
  • Interacts with insect targets that require metal co-factors for function.
Catalog No. 50-004-49210