Learn More
Medchemexpress LLC Abametapir | 1762-34-1 | 184.24 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0045465G
Encompass_Preferred
Abametapir is a metalloproteinase (MMP) inhibitor that targets metalloproteinases crucial for egg hatching and louse development. It can inhibit the hatching of both head and body lice.
  • Targets metalloproteinases
  • Inhibits egg hatching in lice
  • Effective against head and body lice
Catalog No. 50-004-46953