Learn More
Medchemexpress LLC Abametapir | 1762-34-1 | 184.24 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00454650G
Encompass_Preferred
Abametapir is a metalloproteinase (MMP) inhibitor that targets metalloproteinases crucial for egg hatching and louse development, effectively inhibiting the hatching of both head and body lice. It can chelate heavy metal ions such as iron, copper, and zinc, allowing it to interact with various insect targets that require metal co-factors for their function.
  • Targets metalloproteinases critical for egg hatching and louse development
  • Inhibits hatching of both head and body lice
  • Chelates heavy metal ions like iron, copper, and zinc
  • Interacts with metal co-factor dependent targets, including metalloproteinases
Catalog No. 50-004-48775