Learn More
Apexbio Technology LLC Abametapir 1762-34-1 500mg
SDP
Supplier:  Apexbio Technology LLC A4646500
Encompass_Preferred
Abametapir (CAS 1762-34-1) is a small-molecule inhibitor targeting metalloproteinases It is designed to inhibit metalloproteinase enzymes involved in the egg hatching and survival processes of lice Abametapir exerts its biological activity primarily through inhibition of metalloproteinases In in vitro assays abametapir demonstrates ovicidal activity with reported IC50 values approximately in the low micromolar range Based on these pharmacological properties abametapir holds research potential in the management of head lice infestations and laboratory studies on ectoparasite enzyme inhibition and resistance development
Catalog No. 50-000-93686