Learn More
Selleck Chemical LLC Abarelix P1142-5MG
SDP
Supplier:  Selleck Chemical LLC P11425MG
Encompass_Preferred
Abarelix (R3827 PPI 149) is a potent gonadotrophin-releasing hormone (GnRH) antagonist used for prostate cancer treatment
Catalog No. 50-000-43620