Learn More
Medchemexpress LLC HY-13534A 5mg Medchemexpress, Abarelix (Acetate) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13534A5MG
Encompass_Preferred
Medchemexpress, HY-13534A 5mg Abarelix (Acetate) CAS: Abarelix Acetate (PPI 149 Acetate; R 3827 Acetate) is a potent gonadotrophin-releasing hormone (GnRH) antagonist, used for prostate cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9874