Learn More
Cayman Chemical AbarelIxacetate 10mg
SDP
Supplier:  Cayman Chemical 2874610 MG
Encompass_Preferred
A synthetic GnRHR antagonist (IC50 = 3.5 nM); decreases plasma LH levels six hours post-treatment in castrated rats, with levels returning to baseline within 24 hour at 200 μg/kg; transiently decreases plasma testosterone levels in intact rats, with levels returning to baseline within seven days post-treatment at 2 mg/kg
Catalog No. 50-293-9395