Learn More
Medchemexpress LLC HY-108829 1mg Medchemexpress, Abatacept CAS:332348-12-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1088291MG
Encompass_Preferred
Medchemexpress, HY-108829 1mg Abatacept CAS:332348-12-6 Abatacept (CTLA4lg) is a soluble fusion protein consisting of the extra-cellular domain of human CTLA4 and a fragment of the Fc portion of human IgG1 (hinge and CH2 and 3 domains). Abatacept is a selective T-cell co-stimulation modulator and a protein drug for the autoimmune diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8629