Learn More
Alkali Scientific Abbott AxSym Compatible Sample Cups –Polystyrene, 17.5×53mm, 400/Pk
SDP
Supplier:  Alkali Scientific ASCABBOTAXSYM
Encompass_Preferred
Polystyrene Sample Cup, 17.5 × 53 mm — For Abbott AxSym Analyzers. These precision molded sample cups are designed for reliable use with Abbott AxSym analyzers, supporting consistent and accurate sample processing in automated laboratory workflows. With exact 17.5 × 53 mm dimensions, they fit securely into AxSym sample holders and carousel systems, helping prevent alignment issues and reducing the risk of instrument errors. Manufactured from clear medical grade polystyrene, each cup provides excellent optical transparency for easy visual inspection of sample clarity and volume. The rigid, smooth wall construction supports accurate pipetting, consistent sample delivery, and dependable analyzer performance. Disposable cups are non pyrogenic and RNase/DNase free, making them suitable for clinical chemistry and immunoassay testing environments.
Catalog No. 50-006-62606